4-Bromo-N'-methyl-1H-pyrrole-2-carbohydrazide, tech

Lieferant: Apollo Scientific

OR32157-1G
APOSOR32157-1GEA 1570 EUR
APOSOR32157-1G
4-Bromo-N'-methyl-1H-pyrrole-2-carbohydrazide, tech
4-Bromo-N'-methyl-1H-pyrrole-2-carbohydrazide
Formel: C₆H₈BrN₃O
Molecular Weight: 218,051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08281823
CAS-Nummer: 923676-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR