1-(4-Bromo-1H-pyrrole-2-carbonyl)piperidine 95%
Lieferant: Apollo Scientific
OR32158-1G
APOSOR32158-1GEA
224
EUR
APOSOR32158-1G
1-(4-Bromo-1H-pyrrole-2-carbonyl)piperidine 95%
1-(4-Bromo-1H-pyrrole-2-carbonyl)piperidine
Formel:
C₁₀H₁₃BrN₂O Molecular Weight: 257.127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021325 CAS-Nummer: 900019-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...