3-Acetyl-6-(tert-butyl)-2(1H)-pyridinone 95%

Lieferant: Apollo Scientific

Synonyme: 3-Acetyl-6-tert-butyl-1,2-dihydropyridin-2-one

OR32168-1G
APOSOR32168-1GEA 1330 EUR
APOSOR32168-1G
3-Acetyl-6-(tert-butyl)-2(1H)-pyridinone 95%
3-Acetyl-6-(tert-butyl)-2(1H)-pyridinone
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056604
CAS-Nummer: 885950-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR