Methyl 5-[(E)-2-(dimethylamino)ethenyl]-3-phenyl-1,2-oxazole-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32171-1G
APOSOR32171-1GEA 1570 EUR
APOSOR32171-1G
Methyl 5-[(E)-2-(dimethylamino)ethenyl]-3-phenyl-1,2-oxazole-4-carboxylate, tech
Methyl 5-[(E)-2-(dimethylamino)ethenyl]-3-phenyl-1,2-oxazole-4-carboxylate
Formel: C₁₅H₁₆N₂O₃
Molecular Weight: 272,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127101
CAS-Nummer: 1610688-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR