Methyl 1-oxo-2-phenyl-1,2-dihydroisoquinoline-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32173-1G
APOSOR32173-1GEA 1570 EUR
APOSOR32173-1G
Methyl 1-oxo-2-phenyl-1,2-dihydroisoquinoline-4-carboxylate, tech
Methyl 1-oxo-2-phenyl-1,2-dihydroisoquinoline-4-carboxylate
Formel: C₁₇H₁₃NO₃
Molecular Weight: 279,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140322
CAS-Nummer: 73349-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR