Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate 95%

Lieferant: Apollo Scientific

OR32174-1G
APOSOR32174-1GEA 671 EUR
APOSOR32174-1G
Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate 95%
Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate
Formel: C₁₇H₁₄N₂O₃
Molecular Weight: 294,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140334
CAS-Nummer: 446276-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR