Methyl 1-phenyl-3-(pyridin-4-yl)-1H-pyrazole-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32179-1G
APOSOR32179-1GEA 1570 EUR
APOSOR32179-1G
Methyl 1-phenyl-3-(pyridin-4-yl)-1H-pyrazole-4-carboxylate, tech
Methyl 1-phenyl-3-(pyridin-4-yl)-1H-pyrazole-4-carboxylate
Formel: C₁₆H₁₃N₃O₂
Molecular Weight: 279,293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01936047
CAS-Nummer: 400074-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR