Methyl (2E)-2-(3-chlorophenyl)-3-(dimethylamino)prop-2-enoate, tech
Lieferant: Apollo Scientific
OR32182-1G
APOSOR32182-1GEA
1570
EUR
APOSOR32182-1G
Methyl (2E)-2-(3-chlorophenyl)-3-(dimethylamino)prop-2-enoate, tech
Methyl (2E)-2-(3-chlorophenyl)-3-(dimethylamino)prop-2-enoate
Formel:
C₁₂H₁₄ClNO₂ Molecular Weight: 239,698 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04124266 CAS-Nummer: 80510-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...