4-(4-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech

Lieferant: Apollo Scientific

OR32183-1G
APOSOR32183-1GEA 0 EUR
APOSOR32183-1G
4-(4-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech
4-(4-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Formel: C₁₅H₁₁ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00232637
CAS-Nummer: 400075-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR