4'-Phenyl-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,3,4]triazolidine]-5'-one, tech

Lieferant: Apollo Scientific

OR32185-1G
APOSOR32185-1GEA 1570 EUR
APOSOR32185-1G
4'-Phenyl-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,3,4]triazolidine]-5'-one, tech
4'-Phenyl-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,3,4]triazolidine]-5'-one
Formel: C₁₇H₁₇N₃O
Molecular Weight: 279,336 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140495
CAS-Nummer: 131365-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR