5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%

Lieferant: Apollo Scientific

OR32187-1G
APOSOR32187-1GEA 671 EUR
APOSOR32187-1G
5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%
5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
Formel: C₁₂H₁₁ClN₂O₂
Molecular Weight: 250.69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215393
CAS-Nummer: 109925-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR