1-(6-Chlor-3-pyridazinyl)piperidin-4-on 95%

Lieferant: Apollo Scientific

Synonyme: 1-(6-Chloropyridazin-3-yl)piperidin-4-one

OR32188-5G
APOSOR32188-5GEA 180 EUR
APOSOR32188-5G
1-(6-Chlor-3-pyridazinyl)piperidin-4-on 95%
1-(6-Chlor-3-pyridazinyl)piperidin-4-on
Formel: C₉H₁₀ClN₃O
Molecular Weight: 211.648 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665150
CAS-Nummer: 303149-95-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR