Methyl 3-[4-(4-methoxyphenyl)piperazin-1-yl]-3-oxopropanoate, tech
Lieferant: Apollo Scientific
OR32190-1G
APOSOR32190-1GEA
0
EUR
APOSOR32190-1G
Methyl 3-[4-(4-methoxyphenyl)piperazin-1-yl]-3-oxopropanoate, tech
Methyl 3-[4-(4-methoxyphenyl)piperazin-1-yl]-3-oxopropanoate
Formel:
C₁₅H₂₀N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664721 CAS-Nummer: 303151-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...