N-[2-(1H-Pyrrol-1-yl)ethyl]acetamide 95%

Lieferant: Apollo Scientific

OR32191-1G
APOSOR32191-1GEA 1330 EUR
APOSOR32191-1G
N-[2-(1H-Pyrrol-1-yl)ethyl]acetamide 95%
N-[2-(1H-Pyrrol-1-yl)ethyl]acetamide
Formel: C₈H₁₂N₂O
Molecular Weight: 152,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00180783
CAS-Nummer: 73627-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR