Methyl 1-(pyrimidin-2-yl)piperidine-4-carboxylate, tech
Lieferant: Apollo Scientific
OR32195-1G
APOSOR32195-1GEA
1570
EUR
APOSOR32195-1G
Methyl 1-(pyrimidin-2-yl)piperidine-4-carboxylate, tech
Methyl 1-(pyrimidin-2-yl)piperidine-4-carboxylate
Formel:
C₁₁H₁₅N₃O₂ Molecular Weight: 221,256 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664472 CAS-Nummer: 303144-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...