2-Benzoyl-1H-imidazole 95%

Lieferant: Apollo Scientific

OR32196-1G
APOSOR32196-1GEA 224 EUR
APOSOR32196-1G
2-Benzoyl-1H-imidazole 95%
2-Benzoyl-1H-imidazole
Formel: C₁₀H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00707818
CAS-Nummer: 38353-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR