3,5-Dichloro-N-methylbenzamide 95%

Lieferant: Apollo Scientific

OR32199-1G
APOSOR32199-1GEA 427 EUR
APOSOR32199-1G
3,5-Dichloro-N-methylbenzamide 95%
3,5-Dichloro-N-methylbenzamide
Formel: C₈H₇Cl₂NO
Molecular Weight: 204,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243816
CAS-Nummer: 33244-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR