1-Phenylcyclopropancarbonsäure

Lieferant: Apollo Scientific

OR322012-1G
APOSOR322012-1GEA 0 EUR
APOSOR322012-1G
1-Phenylcyclopropancarbonsäure
1-Phenylcyclopropanecarboxylic acid
Formel: C₁₀H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00001288
CAS-Nummer: 6120-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR