6-[(E)-2-[4-(Propan-2-yl)phenyl]ethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32202-1G
APOSOR32202-1GEA 1570 EUR
APOSOR32202-1G
6-[(E)-2-[4-(Propan-2-yl)phenyl]ethenyl]-2,3,4,5-tetrahydropyridazin-3-one, tech
6-[(E)-2-[4-(Propan-2-yl)phenyl]ethenyl]-2,3,4,5-tetrahydropyridazin-3-one
Formel: C₁₅H₁₈N₂O
Molecular Weight: 242,316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793320
CAS-Nummer: 303146-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR