2-Isoxazol-5-yl-4-methylphenol

Lieferant: Apollo Scientific

OR322023-1G
APOSOR322023-1GEA 0 EUR
APOSOR322023-1G
2-Isoxazol-5-yl-4-methylphenol
2-Isoxazol-5-yl-4-methylphenol
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00218657
CAS-Nummer: 164171-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR