Comanic acid

Lieferant: Apollo Scientific

OR322025-250MG
APOSOR322025-250MGEA 0 EUR
APOSOR322025-250MG
Comanic acid
Comanic acid
Formel: C₆H₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00059958
CAS-Nummer: 499-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR