5-Acetyl-6-methyl-2-(piperidin-1-yl)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32204-1G
APOSOR32204-1GEA 1570 EUR
APOSOR32204-1G
5-Acetyl-6-methyl-2-(piperidin-1-yl)pyridine-3-carbonitrile, tech
5-Acetyl-6-methyl-2-(piperidin-1-yl)pyridine-3-carbonitrile
Formel: C₁₄H₁₇N₃O
Molecular Weight: 243,304 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793588
CAS-Nummer: 121348-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR