1,2-Diamino-9,10-anthrachinon

Lieferant: Apollo Scientific
Achtung

OR322042-5G
APOSOR322042-5GEA 0 EUR
APOSOR322042-5G
1,2-Diamino-9,10-anthrachinon
1,2-Diamino-9,10-anthrachinon
Formel: C₁₄H₁₀N₂O₂
Schmelzpunkt: 288 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001219
CAS-Nummer: 1758-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR