N-[(Benzyloxy)carbonyl]serin 

Lieferant: Apollo Scientific

Synonyme: N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine

OR322046-5G
APOSOR322046-5GEA 49.3 EUR
APOSOR322046-5G
N-[(Benzyloxy)carbonyl]serin 
N-Cbz-L-serine
Formel: C₁₁H₁₃NO₅
Molecular Weight: 239,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002662
CAS-Nummer: 1145-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR