1,8-Dinitro-9,10-anthrachinon 95%

Lieferant: Apollo Scientific
Achtung

OR322049-5G
APOSOR322049-5GEA 21.9 EUR
APOSOR322049-5G
1,8-Dinitro-9,10-anthrachinon 95%
1,8-Dinitro-9,10-anthrachinon
Formel: C₁₄H₆N₂O₆
Lagertemperatur: Raumtemperatur
CAS-Nummer: 129-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR