[8-Methoxy-4-(methylamino)-3,4-dihydro-2H-chromen-3-yl]methanol hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: [8-Methoxy-4-(methylamino)-3,4-dihydro-2H-1-benzopyran-3-yl]methanol hydrochloride

OR32205-1G
APOSOR32205-1GEA 1070 EUR
APOSOR32205-1G
[8-Methoxy-4-(methylamino)-3,4-dihydro-2H-chromen-3-yl]methanol hydrochloride 95%
[8-Methoxy-4-(methylamino)-3,4-dihydro-2H-chromen-3-yl]methanol hydrochloride
Formel: C₁₂H₁₈ClNO₃
Molecular Weight: 259,74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663791
CAS-Nummer: 321391-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR