N-Cyclohexylethanolamine

Lieferant: Apollo Scientific

Synonyme: 2-(Cyclohexylamino)ethanol, N-(2-Hydroxyethyl)cyclohexylamine

OR322052-25G
APOSOR322052-25GEA 66.7 EUR
APOSOR322052-25G
N-Cyclohexylethanolamine
N-Cyclohexylethanolamine
Formel: C₈H₁₇NO
Molecular Weight: 143.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021326
CAS-Nummer: 2842-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR