(1-Amino-2,3-dihydro-1H-benzo[f]chromen-2-yl)methanol hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: {1-Amino-1H,2H,3H-naphtho[2,1-b]pyran-2-yl}methanol hydrochloride

OR32206-1G
APOSOR32206-1GEA 1070 EUR
APOSOR32206-1G
(1-Amino-2,3-dihydro-1H-benzo[f]chromen-2-yl)methanol hydrochloride 95%
(1-Amino-2,3-dihydro-1H-benzo[f]chromen-2-yl)methanol hydrochloride
Formel: C₁₄H₁₆ClNO₂
Molecular Weight: 265,74 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663795
CAS-Nummer: 321392-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR