2-(Ethylthio)Ethylamine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 2-(Ethylthio)ethanamine hydrochloride, 2-(Ethylthio)ethanamine hydrochloride(Flake or chunks or pellets)

OR322060-250MG
APOSOR322060-250MGEA 49.3 EUR
APOSOR322060-250MG
2-(Ethylthio)Ethylamine hydrochloride
2-(Ethylthio)Ethylamine hydrochloride
Formel: C₄H₁₁NS·ClH
Molecular Weight: 141,663 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012903
CAS-Nummer: 54303-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR