1-Methylimidazol-2-carbaldehyd
Lieferant: Apollo Scientific
Synonyme:
1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde
OR322067-1G
APOSOR322067-1GEA
0
EUR
APOSOR322067-1G
1-Methylimidazol-2-carbaldehyd
1-Methylimidazol-2-carbaldehyd
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321308 CAS-Nummer: 13750-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...