4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one 95%

Lieferant: Apollo Scientific

OR32207-1G
APOSOR32207-1GEA 1120 EUR
APOSOR32207-1G
4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one 95%
4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one
Formel: C₆H₆Cl₂N₂O
Molecular Weight: 193,031 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793799
CAS-Nummer: 33098-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR