2-(3,4-Dichlorphenyl)ethanol
Lieferant: Apollo Scientific
OR322080-1G
APOSOR322080-1GEA
0
EUR
APOSOR322080-1G
2-(3,4-Dichlorphenyl)ethanol
2-(3,4-Dichlorophenyl)ethanol
Formel:
C₈H₈Cl₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800673 CAS-Nummer: 35364-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...