(3Z)-1-Methyl-3-(2-phenylhydrazin-1-ylidene)-2,3-dihydro-1H-indol-2-one, tech
Lieferant: Apollo Scientific
OR32209-1G
APOSOR32209-1GEA
1570
EUR
APOSOR32209-1G
(3Z)-1-Methyl-3-(2-phenylhydrazin-1-ylidene)-2,3-dihydro-1H-indol-2-one, tech
(3Z)-1-Methyl-3-(2-phenylhydrazin-1-ylidene)-2,3-dihydro-1H-indol-2-one
Formel:
C₁₅H₁₃N₃O Molecular Weight: 251,283 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00194472 CAS-Nummer: 15096-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...