(3E)-3-[2-(4-Methoxyphenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one, tech
Lieferant: Apollo Scientific
OR32210-1G
APOSOR32210-1GEA
1570
EUR
APOSOR32210-1G
(3E)-3-[2-(4-Methoxyphenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one, tech
(3E)-3-[2-(4-Methoxyphenyl)hydrazin-1-ylidene]-1-methyl-2,3-dihydro-1H-indol-2-one
Formel:
C₁₆H₁₅N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01314765 CAS-Nummer: 303984-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...