N-Methyl-1,3-propandiamin 98%

Lieferant: Apollo Scientific

Synonyme: N-Methylpropane-1,3-diamine, 3-Methylamino propylamine

OR322100-5G
APOSOR322100-5GEA 21.9 EUR
APOSOR322100-5G
N-Methyl-1,3-propandiamin 98%
N-Methyl-1,3-propandiamin
Formel: C₄H₁₂N₂
Molecular Weight: 88,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008209
CAS-Nummer: 6291-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR