tert-Butyl-N-hydroxycarbamat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine
OR322101-100G
OR322101-25G
OR322101-500G
APOSOR322101-100GEA
72.5
EUR
APOSOR322101-100G
APOSOR322101-25G
APOSOR322101-500G
tert-Butyl-N-hydroxycarbamat
tert-Butyl-N-hydroxycarbamat
Formel:
C₅H₁₁NO₃ Molecular Weight: 133.15 g/mol Schmelzpunkt: 55…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002107 CAS-Nummer: 36016-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...