tert-Butyl-N-hydroxycarbamat

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine

OR322101-100G OR322101-25G OR322101-500G
APOSOR322101-100GEA 72.5 EUR
APOSOR322101-100G APOSOR322101-25G APOSOR322101-500G
tert-Butyl-N-hydroxycarbamat
tert-Butyl-N-hydroxycarbamat
Formel: C₅H₁₁NO₃
Molecular Weight: 133.15 g/mol
Schmelzpunkt: 55…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002107
CAS-Nummer: 36016-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR