(R)-(-)-N-Benzyl-2-phenylglycinol
Lieferant: Apollo Scientific
Synonyme:
(R)-2-(Benzylamino)-2-phenylethanol
OR322107-1G
APOSOR322107-1GEA
29
EUR
APOSOR322107-1G
(R)-(-)-N-Benzyl-2-phenylglycinol
(R)-(-)-N-Benzyl-2-phenylglycinol
Formel:
C₁₅H₁₇NO Molecular Weight: 227,302 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674035 CAS-Nummer: 14231-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...