(S)-(-)-2-Acetoxypropionic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR322114-1G
OR322114-5G
APOSOR322114-1GEA
0
EUR
APOSOR322114-1G
APOSOR322114-5G
(S)-(-)-2-Acetoxypropionic acid
(S)-(-)-2-Acetoxypropionic acid
Formel:
C₅H₈O₄ Siedepunkt: 142 °C/20mmHg Dichte: 1,18 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191370 CAS-Nummer: 6034-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...