(R)-(+)-1-(p-Tolyl)ethylamine

Lieferant: Apollo Scientific

OR322115-1G
APOSOR322115-1GEA 0 EUR
APOSOR322115-1G
(R)-(+)-1-(p-Tolyl)ethylamine
(R)-(+)-1-(p-Tolyl)ethylamine
Formel: C₉H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00145202
CAS-Nummer: 4187-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR