2-Formyl-2-phenylacetonitrile
Lieferant: Apollo Scientific
OR322124-1G
APOSOR322124-1GEA
132
EUR
APOSOR322124-1G
2-Formyl-2-phenylacetonitrile
2-Formyl-2-phenylacetonitrile
Formel:
C₉H₇NO Molecular Weight: 145,158 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024158 CAS-Nummer: 5841-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...