6-Bromohexanamide

Lieferant: Apollo Scientific

OR322129-5G
APOSOR322129-5GEA 45.1 EUR
APOSOR322129-5G
6-Bromohexanamide
6-Bromohexanamide
Formel: C₆H₁₂BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00236597
CAS-Nummer: 57817-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR