1-Benzoylnaphthalin
Lieferant: Apollo Scientific
Synonyme:
1-Naphthyl phenyl ketone
OR322138-1G
APOSOR322138-1GEA
212
EUR
APOSOR322138-1G
1-Benzoylnaphthalin
1-Benzoylnaphthalin
Formel:
C₁₇H₁₂O Molecular Weight: 232,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046431 CAS-Nummer: 642-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...