N-[(3,5-Dimethyl-1,2-oxazol-4-yl)methyl]aniline 95%

Lieferant: Apollo Scientific

OR32214-1G
APOSOR32214-1GEA 339 EUR
APOSOR32214-1G
N-[(3,5-Dimethyl-1,2-oxazol-4-yl)methyl]aniline 95%
N-[(3,5-Dimethyl-1,2-oxazol-4-yl)methyl]aniline
Formel: C₁₂H₁₄N₂O
Molecular Weight: 202.252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124300
CAS-Nummer: 733796-14-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR