3-Brom-N,N-dimethylbenzamid
Lieferant: Apollo Scientific
OR322148-250MG
APOSOR322148-250MGEA
30.6
EUR
APOSOR322148-250MG
3-Brom-N,N-dimethylbenzamid
3-Brom-N,N-dimethylbenzamid
Formel:
C₉H₁₀BrNO Molecular Weight: 228,0858 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01215179 CAS-Nummer: 24167-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...