(2Z)-2-[4-(3,3-Dimethyl-2-oxoazetidin-1-yl)phenyl]-3-(dimethylamino)prop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR32215-1G
APOSOR32215-1GEA 1570 EUR
APOSOR32215-1G
(2Z)-2-[4-(3,3-Dimethyl-2-oxoazetidin-1-yl)phenyl]-3-(dimethylamino)prop-2-enenitrile, tech
(2Z)-2-[4-(3,3-Dimethyl-2-oxoazetidin-1-yl)phenyl]-3-(dimethylamino)prop-2-enenitrile
Formel: C₁₆H₁₉N₃O
Molecular Weight: 269,342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793385
CAS-Nummer: 303985-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR