(3-Brompropyl)ammoniumbromid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Amino-3-brompropan Hydrobromid, 3-Brompropylamin Hydrobromid

OR322151-100G OR322151-500G OR322151-25G
APOSOR322151-100GEA 31.9 EUR
APOSOR322151-100G APOSOR322151-500G APOSOR322151-25G
(3-Brompropyl)ammoniumbromid
3-Brompropylamine hydrobromide
Formel: BrH₂CCH₂CH₂NH₃Br
Molecular Weight: 218.92 g/mol
Schmelzpunkt: 171…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012912
CAS-Nummer: 5003-71-4
EINECS: 225-675-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR