3-Chloro-N-(3-hydroxyphenyl)-2,2-dimethylpropanamide, tech
Lieferant: Apollo Scientific
OR32216-1G
APOSOR32216-1GEA
1570
EUR
APOSOR32216-1G
3-Chloro-N-(3-hydroxyphenyl)-2,2-dimethylpropanamide, tech
3-Chloro-N-(3-hydroxyphenyl)-2,2-dimethylpropanamide
Formel:
C₁₁H₁₄ClNO₂ Molecular Weight: 227,687 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793397 CAS-Nummer: 303986-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...