4-(Methylthio)benzylamin

Lieferant: Apollo Scientific

OR322186-1G OR322186-5G
APOSOR322186-1GEA 0 EUR
APOSOR322186-1G APOSOR322186-5G
4-(Methylthio)benzylamin
4-(Methylthio)benzylamin
Formel: C₈H₁₁NS
Lagertemperatur: Raumtemperatur
MDL: MFCD01310831
CAS-Nummer: 83171-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR