(2-Aminophenyl)acetonitril

Lieferant: Apollo Scientific

Synonyme: 2-(2-Aminophenyl)acetonitrile, 2-Aminobenzyl cyanide, (2-Aminophenyl)acetonitrile

OR322191-250MG
APOSOR322191-250MGEA 53.8 EUR
APOSOR322191-250MG
(2-Aminophenyl)acetonitril
(2-Aminophenyl)acetonitril
Formel: C₈H₈N₂
Molecular Weight: 132,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00091082
CAS-Nummer: 2973-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR